C12H316CF586 Chemcia Scientific, LLC

1-(2-Amino-5-bromo-3-iodo-phenyl)-ethanone

  • Cas Number : 1985633-67-7
  • Molecular Weight : 339.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-amino-5-bromo-3-iodophenyl)ethan-1-one
Inchi Code 1S/C8H7BrINO/c1-4(12)6-2-5(9)3-7(10)8(6)11/h2-3H,11H2,1H3
Inchi Key LBKOALQXKQERGF-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=11200

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11200
Availability details for
Quantity
  • Availability
  • Add to Cart