C12H316CF58A Chemcia Scientific, LLC

(3-Nitro-5-trifluoromethyl-phenyl)-methanol

  • MDL number MFCD11040241
  • Cas Number : 180146-66-1
  • Molecular Weight : 221.13712

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11204

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11204
Availability details for
Quantity
  • Availability
  • Add to Cart