C12H316CF597 Chemcia Scientific, LLC

3-Chloro-5-fluoro-2H-indazole

  • MDL number MFCD28540322
  • Molecular Weight : 170.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-5-fluoro-2H-indazole
Inchi Code 1S/C7H4ClFN2/c8-7-5-3-4(9)1-2-6(5)10-11-7/h1-3H,(H,10,11)
Inchi Key QCTONHPGUDYART-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=11318

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11318
Availability details for
Quantity
  • Availability
  • Add to Cart