C12H316CF599 Chemcia Scientific, LLC

5-Bromo-2-iodo-3-nitro-benzoic acid methyl ester

  • MDL number MFCD12172981
  • Cas Number : 855749-51-8
  • Molecular Weight : 385.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 5-bromo-2-iodo-3-nitrobenzoate
Inchi Code 1S/C8H5BrINO4/c1-15-8(12)5-2-4(9)3-6(7(5)10)11(13)14/h2-3H,1H3
Inchi Key STOFEGROEWKXTB-UHFFFAOYSA-N

Description

Purity 90%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=11339

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11339
Availability details for
Quantity
  • Availability
  • Add to Cart