C12H316CF59C Chemcia Scientific, LLC

2-Acetylamino-5-chloro-3-methyl-benzoic acid methyl ester

  • Molecular Weight : 241.67619

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11596

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11596
Availability details for
Quantity
  • Availability
  • Add to Cart