C12H316CF59F Chemcia Scientific, LLC

(5-Bromo-1-methyl-1H-pyrazol-4-yl)-methanol

  • MDL number MFCD28140435
  • Cas Number : 1415638-13-9
  • Molecular Weight : 191.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-bromo-1-methyl-1H-pyrazol-4-yl)methanol
Inchi Code 1S/C5H7BrN2O/c1-8-5(6)4(3-9)2-7-8/h2,9H,3H2,1H3
Inchi Key DYXXSYMFUPNKBH-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=11393

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11393
Availability details for
Quantity
  • Availability
  • Add to Cart