C12H316CF5A5 Chemcia Scientific, LLC

3-Amino-2-bromo-6-fluoro-benzoic acid methyl ester

  • MDL number MFCD22570415
  • Cas Number : 1446261-65-9
  • Molecular Weight : 248.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-amino-2-bromo-6-fluorobenzoate
Inchi Code 1S/C8H7BrFNO2/c1-13-8(12)6-4(10)2-3-5(11)7(6)9/h2-3H,11H2,1H3
Inchi Key BEGPPSBHXZMUGJ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=11504

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11504
Availability details for
Quantity
  • Availability
  • Add to Cart