C12H316CF5A6 Chemcia Scientific, LLC

2-Bromo-6-fluoro-3-nitro-benzoic acid methyl ester

  • MDL number MFCD22570414
  • Cas Number : 1610060-53-1
  • Molecular Weight : 278.03575

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11505

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11505
Availability details for
Quantity
  • Availability
  • Add to Cart