C12H316CF5A7 Chemcia Scientific, LLC

(1,4,6,7-Tetrahydro-pyrano[4,3-c]pyrazol-3-yl)-methanol

  • MDL number MFCD20491781
  • Cas Number : 1351386-56-5
  • Molecular Weight : 154.16995

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11546

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11546
Availability details for
Quantity
  • Availability
  • Add to Cart