C12H316CF5AD Chemcia Scientific, LLC

5-Chloro-2H-indazole-3-carboxylic acid

  • Molecular Weight : 196.59425

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11599

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11599
Availability details for
Quantity
  • Availability
  • Add to Cart