C12H316CF5B0 Chemcia Scientific, LLC

(2-Amino-5-chloro-3-methyl-phenyl)-methanol

  • MDL number MFCD20702609
  • Cas Number : 775331-65-2
  • Molecular Weight : 171.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-amino-5-chloro-3-methylphenyl)methanol
Inchi Code 1S/C8H10ClNO/c1-5-2-7(9)3-6(4-11)8(5)10/h2-3,11H,4,10H2,1H3
Inchi Key UREQNFKFFMSGSZ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=11601

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11601
Availability details for
Quantity
  • Availability
  • Add to Cart