C12H316CF5B1 Chemcia Scientific, LLC

N-(2-Methoxy-4-methyl-pyridin-3-yl)-acetamide

  • Cas Number : 76005-98-6
  • Molecular Weight : 180.20819

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11602

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11602
Availability details for
Quantity
  • Availability
  • Add to Cart