C12H316CF5B2 Chemcia Scientific, LLC

2-Bromo-5-fluoro-pyridine 1-oxide

  • MDL number MFCD09261236
  • Cas Number : 935534-39-7
  • Molecular Weight : 191.98816

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11603

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11603
Availability details for
Quantity
  • Availability
  • Add to Cart