C12H316CF5B4 Chemcia Scientific, LLC

6-Fluoro-1H-pyrrolo[3,2-b]pyridin-3-ylamine

  • MDL number MFCD12963041
  • Cas Number : 1190319-66-4
  • Molecular Weight : 151.14437

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11621

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11621
Availability details for
Quantity
  • Availability
  • Add to Cart