C12H316CF5B6 Chemcia Scientific, LLC

4-Fluoro-2-methoxy-5-nitro-benzamide

  • Cas Number : 1569987-64-9
  • Molecular Weight : 214.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-fluoro-2-methoxy-5-nitrobenzamide

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=11681

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11681
Availability details for
Quantity
  • Availability
  • Add to Cart