C12H316CF5B9 Chemcia Scientific, LLC

2-Amino-3-bromo-5-nitro-benzoic acid

  • Cas Number : 773108-00-2
  • Molecular Weight : 261.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-amino-3-bromo-5-nitrobenzoic acid
Inchi Code 1S/C7H5BrN2O4/c8-5-2-3(10(13)14)1-4(6(5)9)7(11)12/h1-2H,9H2,(H,11,12)
Inchi Key MDXOSJZNEXCDOQ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=11780

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11780
Availability details for
Quantity
  • Availability
  • Add to Cart