C12H316CF5BD Chemcia Scientific, LLC

5-Iodo-benzo[d]isoxazol-3-ylamine

  • MDL number MFCD11847106
  • Cas Number : 1260812-04-1
  • Molecular Weight : 260.0351

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11835

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11835
Availability details for
Quantity
  • Availability
  • Add to Cart