C12H316CF5C2 Chemcia Scientific, LLC

3-Bromo-2-bromomethyl-benzoic acid

  • MDL number MFCD11847530
  • Cas Number : 1194354-14-7
  • Molecular Weight : 293.94382

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11915

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11915
Availability details for
Quantity
  • Availability
  • Add to Cart