C12H316CF5C8 Chemcia Scientific, LLC

6-Chloro-3-fluoro-pyridine-2-carbonitrile

  • MDL number MFCD18250470
  • Cas Number : 1207609-52-6
  • Molecular Weight : 156.54764

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11607

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11607
Availability details for
Quantity
  • Availability
  • Add to Cart