C12H316CF5CF Chemcia Scientific, LLC

8-Bromo-6-fluoro-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester

  • MDL number MFCD23115410
  • Cas Number : 1421312-22-2
  • Molecular Weight : 287.08986

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=12185

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12185
Availability details for
Quantity
  • Availability
  • Add to Cart