C12H316CF5D0 Chemcia Scientific, LLC

3-Fluoro-1-oxy-pyridine-2-carbonitrile

  • Cas Number : 1344734-69-5
  • Molecular Weight : 138.10201

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=12186

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12186
Availability details for
Quantity
  • Availability
  • Add to Cart