C12H316CF5D4 Chemcia Scientific, LLC

4-Chloro-2-fluoro-5-hydroxy-benzaldehyde

  • MDL number MFCD26687878
  • Cas Number : 1507007-33-1
  • Molecular Weight : 174.56013

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=12075

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12075
Availability details for
Quantity
  • Availability
  • Add to Cart