C12H316CF5D6 Chemcia Scientific, LLC

2-Fluoro-5-methoxy-4-nitro-benzoic acid

  • MDL number MFCD26395470
  • Cas Number : 1001345-80-7
  • Molecular Weight : 215.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-fluoro-5-methoxy-4-nitrobenzoic acid
Inchi Code 1S/C8H6FNO5/c1-15-7-2-4(8(11)12)5(9)3-6(7)10(13)14/h2-3H,1H3,(H,11,12)
Inchi Key JXCYBTVSPGQRCM-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=12101

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12101
Availability details for
Quantity
  • Availability
  • Add to Cart