C12H316CF5DC Chemcia Scientific, LLC

2H-Pyrazolo[3,4-b]pyridin-3-ylamine

  • MDL number MFCD13176002
  • Cas Number : 64781-70-0
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2H-pyrazolo[3,4-b]pyridin-3-amine
Inchi Code 1S/C6H6N4/c7-5-4-2-1-3-8-6(4)10-9-5/h1-3H,(H3,7,8,9,10)
Inchi Key LUAQTQAFUORQHV-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=12178

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12178
Availability details for
Quantity
  • Availability
  • Add to Cart