C12H316CF5E1 Chemcia Scientific, LLC

5-Chloro-2-hydroxy-4-methoxy-benzoic acid

  • Cas Number : 1378866-39-7
  • Molecular Weight : 202.59559

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=12189

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12189
Availability details for
Quantity
  • Availability
  • Add to Cart