C12H316CF5EC Chemcia Scientific, LLC

6-Chloro-2-methoxy-pyridine-3-sulfonyl chloride

  • MDL number MFCD17014998
  • Cas Number : 1261663-47-1
  • Molecular Weight : 242.08165

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=12200

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12200
Availability details for
Quantity
  • Availability
  • Add to Cart