C12H316CF5F1 Chemcia Scientific, LLC

5-Bromo-2-iodo-4-methyl-benzoic acid

  • Cas Number : 2091451-28-2
  • Molecular Weight : 340.94422

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=12337

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12337
Availability details for
Quantity
  • Availability
  • Add to Cart