C12H316CF5F5 Chemcia Scientific, LLC

(3,4-Difluoro-phenyl)-dimethyl-amine

  • Cas Number : 933585-36-5
  • Molecular Weight : 157.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,4-difluoro-N,N-dimethylaniline
Inchi Code 1S/C8H9F2N/c1-11(2)6-3-4-7(9)8(10)5-6/h3-5H,1-2H3
Inchi Key KCGSFIPMUFBOPM-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=12201

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12201
Availability details for
Quantity
  • Availability
  • Add to Cart