C12H316CF5FA Chemcia Scientific, LLC

(2-Amino-3-bromo-4-methyl-phenyl)-methanol

  • Molecular Weight : 216.079

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=12436

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12436
Availability details for
Quantity
  • Availability
  • Add to Cart