C12H316CF5FB Chemcia Scientific, LLC

2-Amino-3-bromo-4-methyl-benzaldehyde

  • Molecular Weight : 214.06306

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=12437

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12437
Availability details for
Quantity
  • Availability
  • Add to Cart