C12H316CF5FD Chemcia Scientific, LLC

4,5-Dichloro-2-methoxy-benzoic acid methyl ester

  • Cas Number : 1239783-16-4
  • Molecular Weight : 235.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4,5-dichloro-2-methoxybenzoate
Inchi Code 1S/C9H8Cl2O3/c1-13-8-4-7(11)6(10)3-5(8)9(12)14-2/h3-4H,1-2H3
Inchi Key JNYABXQGKADMCQ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=12443

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12443
Availability details for
Quantity
  • Availability
  • Add to Cart