C12H316CF606 Chemcia Scientific, LLC

2-Bromomethyl-4-chloro-5-methoxy-benzoic acid methyl ester

  • Molecular Weight : 293.5464

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=12711

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12711
Availability details for
Quantity
  • Availability
  • Add to Cart