C12H316CF608 Chemcia Scientific, LLC

5-Amino-6-fluoro-pyridine-2-carbonitrile

  • MDL number MFCD28721946
  • Cas Number : 1801842-10-3
  • Molecular Weight : 137.11728

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=12844

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12844
Availability details for
Quantity
  • Availability
  • Add to Cart