C12H316CF60B Chemcia Scientific, LLC

4-Chloro-2,6-diiodo-3-methyl-phenol

  • Cas Number : 2227035-28-9
  • Molecular Weight : 394.3791

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=12980

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12980
Availability details for
Quantity
  • Availability
  • Add to Cart