C12H316CF60E Chemcia Scientific, LLC

6-Chloro-4-oxo-1,4-dihydro-quinoline-3-carbonitrile

  • MDL number MFCD09946359
  • Cas Number : 71083-61-9
  • Molecular Weight : 204.61715

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=13014

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13014
Availability details for
Quantity
  • Availability
  • Add to Cart