C12H316CF610 Chemcia Scientific, LLC

5-Chloro-4-methoxy-pyridin-2-ylamine

  • MDL number MFCD13689017
  • Cas Number : 662117-63-7
  • Molecular Weight : 158.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-4-methoxypyridin-2-amine
Inchi Code 1S/C6H7ClN2O/c1-10-5-2-6(8)9-3-4(5)7/h2-3H,1H3,(H2,8,9)
Inchi Key FXVNVJFNMIOKOL-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=13062

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13062
Availability details for
Quantity
  • Availability
  • Add to Cart