C12H316CF611 Chemcia Scientific, LLC

3-Acetylamino-4-ethyl-benzoic acid methyl ester

  • Molecular Weight : 221.25825

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=13176

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13176
Availability details for
Quantity
  • Availability
  • Add to Cart