C12H316CF612 Chemcia Scientific, LLC

5-Bromo-3-nitro-1H-indole

  • MDL number MFCD15526778
  • Cas Number : 1260655-97-7
  • Molecular Weight : 241.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-3-nitro-1H-indole
Inchi Code 1S/C8H5BrN2O2/c9-5-1-2-7-6(3-5)8(4-10-7)11(12)13/h1-4,10H
Inchi Key OMCFUDPGQYOPBI-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=13182

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13182
Availability details for
Quantity
  • Availability
  • Add to Cart