C12H316CF615 Chemcia Scientific, LLC

4-Amino-3,5-dichloro-2-methyl-benzoic acid methyl ester

  • Molecular Weight : 234.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-amino-3,5-dichloro-2-methylbenzoate
Inchi Code 1S/C9H9Cl2NO2/c1-4-5(9(13)14-2)3-6(10)8(12)7(4)11/h3H,12H2,1-2H3
Inchi Key ZGBCZBDKNGLVCR-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=13392

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13392
Availability details for
Quantity
  • Availability
  • Add to Cart