C12H316CF61C Chemcia Scientific, LLC

6-Chloro-3-fluoro-2-nitro-phenol

  • Cas Number : 872363-74-1
  • Molecular Weight : 191.54711

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=13393

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13393
Availability details for
Quantity
  • Availability
  • Add to Cart