C12H316CF621 Chemcia Scientific, LLC

5-Acetylamino-2-methyl-benzoic acid methyl ester

  • MDL number MFCD00229666
  • Cas Number : 91133-72-1
  • Molecular Weight : 207.23116

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Purple Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=13396

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13396
Availability details for
Quantity
  • Availability
  • Add to Cart