C12H316CF624 Chemcia Scientific, LLC

2-Hydroxymethyl-5-methoxy-benzoic acid

  • MDL number MFCD17676561
  • Cas Number : 647844-37-9
  • Molecular Weight : 182.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(hydroxymethyl)-5-methoxybenzoic acid
Inchi Code 1S/C9H10O4/c1-13-7-3-2-6(5-10)8(4-7)9(11)12/h2-4,10H,5H2,1H3,(H,11,12)
Inchi Key ZJJVNQSJXSXQJV-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=13399

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13399
Availability details for
Quantity
  • Availability
  • Add to Cart