C12H316CF626 Chemcia Scientific, LLC

2-Nitroamino-isonicotinic acid

  • Cas Number : 84249-03-6
  • Molecular Weight : 183.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(nitroamino)isonicotinic acid
Inchi Code 1S/C6H5N3O4/c10-6(11)4-1-2-7-5(3-4)8-9(12)13/h1-3H,(H,7,8)(H,10,11)
Inchi Key RTNWCTSITRLRJK-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=13401

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13401
Availability details for
Quantity
  • Availability
  • Add to Cart