C12H316CF627 Chemcia Scientific, LLC

N-(4-Methyl-5-nitro-pyridin-2-yl)-acetamide

  • MDL number MFCD05662402
  • Cas Number : 90765-02-9
  • Molecular Weight : 195.17923

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=13402

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13402
Availability details for
Quantity
  • Availability
  • Add to Cart