C12H316CF630 Chemcia Scientific, LLC

N-(3-Benzyloxy-5-chloro-pyrazin-2-yl)-acetamide

  • Molecular Weight : 277.71249

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=14829

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14829
Availability details for
Quantity
  • Availability
  • Add to Cart