C12H316CF633 Chemcia Scientific, LLC

1-(2-Amino-5-thiocyanato-phenyl)-ethanone

  • Molecular Weight : 192.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-acetyl-4-aminophenyl thiocyanate
Inchi Code 1S/C9H8N2OS/c1-6(12)8-4-7(13-5-10)2-3-9(8)11/h2-4H,11H2,1H3
Inchi Key DWWHSRQDHPRWIX-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=13697

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13697
Availability details for
Quantity
  • Availability
  • Add to Cart