C12H316CF637 Chemcia Scientific, LLC

5-Fluoro-1H-indazole-3-carboxylic acid methyl ester

  • MDL number MFCD07371576
  • Cas Number : 78155-73-4
  • Molecular Weight : 194.16674

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=13703

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=13703
Availability details for
Quantity
  • Availability
  • Add to Cart