C12H316CF63B Chemcia Scientific, LLC

1-Bromo-2-fluoro-4-methoxy-3-nitro-benzene

  • MDL number MFCD20491421
  • Cas Number : 1352317-80-6
  • Molecular Weight : 250.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-3-fluoro-2-nitrophenyl methyl ether
Inchi Code 1S/C7H5BrFNO3/c1-13-5-3-2-4(8)6(9)7(5)10(11)12/h2-3H,1H3
Inchi Key LJJAOMRFFXQENV-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=14112

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14112
Availability details for
Quantity
  • Availability
  • Add to Cart