C12H316CF63E Chemcia Scientific, LLC

(2-Amino-4-bromo-5-fluoro-phenyl)-methanol

  • Cas Number : 2091283-08-6
  • Molecular Weight : 220.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-amino-4-bromo-5-fluorophenyl)methanol
Inchi Code 1S/C7H7BrFNO/c8-5-2-7(10)4(3-11)1-6(5)9/h1-2,11H,3,10H2
Inchi Key YFCFFDFBFJWQSW-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=14187

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14187
Availability details for
Quantity
  • Availability
  • Add to Cart