C12H316CF641 Chemcia Scientific, LLC

3-Cyano-2-methyl-benzoic acid methyl ester

  • MDL number MFCD14584407
  • Cas Number : 93340-09-1
  • Molecular Weight : 175.18873

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=14190

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14190
Availability details for
Quantity
  • Availability
  • Add to Cart